Merck
Emerging roles of anacardic acid and its derivatives: a pharmacological overview.
Basic & clinical pharmacology & toxicology (2011-11-23)
Mahadevappa Hemshekhar, Martin Sebastin Santhosh, Kempaiah Kemparaju, Kesturu S Girish
RESUMEN
Anacardic acid (AA) is a bioactive phytochemical found in nutshell of Anacardium occidentale. Chemically, it is a mixture of several closely related organic compounds, each consisting of salicylic acid substituted with an alkyl chain. The traditional Ayurveda depicts nutshell oil as a medicinal remedy for alexeritic, amebicidal, gingivitis, malaria and syphilitic ulcers. However, the enduring research and emerging evidence suggests that AA could be a potent target molecule with bactericide, fungicide, insecticide, anti-termite and molluscicide properties and as a therapeutic agent in the treatment of the most serious pathophysiological disorders like cancer, oxidative damage, inflammation and obesity. Furthermore, AA was found to be a common inhibitor of several clinically targeted enzymes such as NFκB kinase, histone acetyltransferase (HATs), lipoxygenase (LOX-1), xanthine oxidase, tyrosinase and ureases. In view of this, we have made an effort to summarize the ongoing research on the therapeutical role of AA and its derivatives. The current MiniReview sheds light on the pharmacological applications, toxicity and allergic responses associated with AA and its derivatives. Although the available records are promising, much more detailed investigations into the therapeutical properties, particularly the anti-cancer and anti-inflammatory activities, are urgently needed. We hope the present MiniReview will attract and encourage further research on elucidating and appreciating the possible curative properties of AA and its derivatives in the management of multifactorial diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anacardic acid