Merck
  • A Surface Site Interaction Point Method for Dissipative Particle Dynamics Parametrization: Application to Alkyl Ethoxylate Surfactant Self-Assembly.
A Surface Site Interaction Point Method for Dissipative Particle Dynamics Parametrization: Application to Alkyl Ethoxylate Surfactant Self-Assembly.
The journal of physical chemistry. B (2020-06-09)
Ennio Lavagnini, Joanne L Cook, Patrick B Warren, Mark J Williamson, Christopher A Hunter
RESUMEN
Dissipative particle dynamics (DPD) is a coarse-grained approach to the simulation of large supramolecular systems, but one limitation has been that the parameters required to describe the noncovalent interactions between beads are not readily accessible. A first-principles computational method has been developed so that bead interaction parameters can be calculated directly from ab initio gas-phase molecular electrostatic potential surfaces of the molecular fragments that represent the beads. A footprinting algorithm converts the molecular electrostatic potential surfaces into a discrete set of surface site interaction points (SSIPs), and these SSIPs are used in the SSIMPLE (surface site interaction model for the properties of liquids at equilibrium) algorithm to calculate the free energies of transfer of one bead into a solution of any other bead. The bead transfer free energies are then converted into the required DPD interaction parameters for all pairwise combinations of different beads. The reliability of the parameters was demonstrated using DPD simulations of a range of alkyl ethoxylate surfactants. The simulations reproduce the experimentally determined values of the critical micelle concentration and mean aggregation number well for all 22 surfactants studied.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tetraethylene glycol monooctyl ether, liquid, ≥98% (GC)