Merck
Impaired Glucose-Stimulated Proinsulin Secretion Is an Early Marker of β-Cell Impairment Before Prediabetes Stage.
The Journal of clinical endocrinology and metabolism (2019-05-11)
Ying Yang, Min Wang, Jingzhi Tong, Zuoliang Dong, Min Deng, Xiaojun Ren, Hui Li, Jing Yang, Zhaowei Meng, Jinhong Sun, Qing He, Ming Liu
RESUMEN
Evidence indicates that there is substantial impairment/loss of β-cell function/mass even before prediabetes. Elevated plasma proinsulin is a sign of β-cell dysfunction in patients with diabetes/prediabetes. However, the dynamic changes of glucose stimulated proinsulin secretion (GSPS) among nondiabetic individuals remain obscure. To examine GSPS and glucose-stimulated insulin secretion (GSIS) among individuals with normal glucose tolerance (NGT) and impaired glucose tolerance (IGT) and to evaluate whether impaired GSPS is an early biomarker of β-cell impairment in individuals with NGT who have subthreshold postprandial plasma glucose (PPG). We evaluated GSPS and GSIS in 116 Chinese adults without diabetes (mean age ± SD, 33.31 ± 9.10 years; mean BMI, 25.24 ± 4.20 kg/m2) with fasting plasma glucose (FPG) < 5.6 mmol/L. Based on 2hPPG, the participants were divided into three groups: NGT1 (2hPPG < 6.67 mmol/L), NGT2 (6.67 ≤ 2hPPG < 7.78 mmol/L), and IGT (7.78 ≤ 2hPPG<11.1 mmol/L). We analyzed the association of GSIS and GSPS with commonly used indexes of β-cell function, insulin resistance and family history of diabetes. Although not diagnosed with prediabetes, the individuals with NGT2 have clinical characteristics and high diabetes risk factors similar to those of the IGT group. However, unlike individuals with IGT, NGT2 participants did not exhibit a delayed GSIS. Instead, GSPS was impaired in NGT2 groups but not in NGT1 group. This study suggests that impaired GSPS, but not impaired GSIS, may serve as an early biomarker to identify a subpopulation of NGT with a high risk of diabetes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Human GSTP1 ELISA Kit, for cell culture supernatants, plasma, and serum samples