Merck
  • Lipoxin A4 antagonizes the mitogenic effects of leukotriene D4 in human renal mesangial cells. Differential activation of MAP kinases through distinct receptors.
Lipoxin A4 antagonizes the mitogenic effects of leukotriene D4 in human renal mesangial cells. Differential activation of MAP kinases through distinct receptors.
The Journal of biological chemistry (2000-06-28)
B McMahon, C Stenson, F McPhillips, A Fanning, H R Brady, C Godson
RESUMEN
The lipoxygenase-derived eicosanoids leukotrienes and lipoxins are well defined regulators of hemeodynamics and leukocyte recruitment in inflammatory conditions. Here, we describe a novel bioaction of lipoxin A(4) (LXA(4)), namely inhibition of leukotriene D(4) (LTD(4))-induced human renal mesangial cell proliferation, and investigate the signal transduction mechanisms involved. LXA(4) blocked LTD(4)-stimulated phosphatidylinositol 3-kinase (PI 3-kinase) activity in parallel to inhibition of LTD(4)-induced mesangial cell proliferation. Screening of a human mesangial cell cDNA library revealed expression of the recently described cys-leukotriene(1)/LTD(4) receptor. LTD(4)-induced mesangial cell proliferation required both extracellular-related signal regulated kinase (erk) and PI 3-kinase activation and may involve platelet-derived growth factor receptor transactivation. LTD(4)-stimulated the MAP kinases erk and p38 via a pertussis toxin (PTX)-sensitive pathway dependent on PI 3-kinase and protein kinase C activation. On screening a cDNA library, mesangial cells were found to express the previously described LXA(4) receptor. In contrast to LTD(4), LXA(4) showed differential activation of erk and p38. LXA(4) activation of erk was insensitive to PTX and PI 3-kinase inhibition, whereas LXA(4) activation of p38 was sensitive to PTX and could be blocked by the LTD(4) receptor antagonist SKF 104353. These data suggest that LXA(4) stimulation of the MAP kinase superfamily involves two distinct receptors: one shared with LTD(4) and coupled to a PTX-sensitive G protein (G(i)) and a second coupled via an alternative G protein, such as G(q) or G(12), to erk activation. These data expand on the spectrum of LXA(4) bioactions within an inflammatory milieu.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Lipoxin A4, Lipoxin A₄, CAS 89663-86-5, is a potent inhibitor of cytotoxic activity of human natural killer cells. Shown to be as potent as LTB4 in stimulating human neutrophils to generate superoxides.