Merck
  • TLC-electrostatic field induced spray ionization-MS analysis of diverse structural skeletons and its coupling with TLC bioautography for characterization of lipase inhibitory components in American ginseng.
TLC-electrostatic field induced spray ionization-MS analysis of diverse structural skeletons and its coupling with TLC bioautography for characterization of lipase inhibitory components in American ginseng.
Journal of pharmaceutical and biomedical analysis (2019-06-24)
Peiliang Zhang, Lei Zhang, Jiyao Shi, Na Zhang, Yue Li, Tao Wu, Zhihong Cheng
RESUMEN
Natural compounds with diverse structural skeletons have different ionization efficiencies and different mass spectrometry (MS) responses. Some key factors influencing the electrostatic field induced spray ionization (EFISI)-MS for a variety of natural compounds have been optimized and improved. Fifteen reference substances representing ten well-known skeletons of natural products including alkaloids, flavonoids, phenolic acids, lignans, coumarins, anthraquinones, monoterpenoids, sesquiterpenoids, diterpenoids and triterpenoids, were selected and investigated for their EFISI-MSn responses on TLC plates using a dot-blot test. The optimized ionization conditions for these compounds in the positive ion mode were achieved, together with their limits of detection. In addition, to avoid the limitation of some compounds being difficultly ionized in the positive ion mode, the negative ion mode of the TLC-EFISI-MS method was developed and optimized for the first time. By coupling with a TLC bioautographic assay, nine lipase inhibitory components in American ginseng (Panax quinquefolium roots) have been successfully identified/ tentatively identified in situ by their EFISI-MSn data and further confirmed by comparisons of their Rf values and MSn data with those of reference substances. These lipase inhibitory compounds were 24(S)-pseudo-ginsenoside F11, ginsenosides Rg1, Re, XVII, Rc, Rb2/Rb3, Rb1, Ro and malonyl-ginsenoside Rb1. This is the first report that the TLC-EFISI-MSn method is adapted to a broad-spectrum analysis of structural skeletons present in herbal medicines.