Merck
  • Sulfide as a confounding factor in toxicity tests with the sea urchin Paracentrotus lividus: comparisons with chemical analysis data.
Sulfide as a confounding factor in toxicity tests with the sea urchin Paracentrotus lividus: comparisons with chemical analysis data.
Environmental toxicology and chemistry (2004-02-26)
Chiara Losso, Alessandra Arizzi Novelli, Marco Picone, Annamaria Volpi Ghirardini, Pier Francesco Ghetti, Danilo Rudello, Paolo Ugo
RESUMEN
Sperm cell and embryo toxicity tests with the sea urchin Paracentrotus lividus were performed to assess the toxicity of sulfide, which is considered a confounding factor in toxicity tests. For improved information on the sensitivity of these methods to sulfide, experiments were performed in the same aerobic conditions used for testing environmental samples, with sulfide concentrations being monitored at the same time by cathodic stripping voltammetry. New toxicity data for sulfide expressed as median effective concentration (EC50) and no-observed-effect concentration (NOEC) are reported. The EC50 value for the embryo toxicity test (total sulfide at 0.43 mg/L) was three times lower than for the sperm cell test (total sulfide at 1.20 mg/L), and the NOEC values were similar (on the order of total sulfide at 10(-1) mg/L) for both tests. The decrease in sulfide concentration during the bioassay as a consequence of possible oxidation of sulfide by dissolved oxygen was determined by voltammetric analysis, indicating a half-life of about 50 min in the presence of gametes. To check the influence of sulfide concentrations on toxicity effects in real samples, toxicity (with the sperm cell toxicity test) and chemical analyses also were performed in pore-water samples collected with an in situ sampler in sediments of the Lagoon of Venice (Italy). A highly positive correlation between increased acute toxicity and increased sulfide concentration was found. Examination of data revealed that sulfide is a real confounding factor in toxicity testing in anoxic environmental samples containing concentrations above the sensitivity limit of the method.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
RIPAb+ SMN - RIP Validated Antibody and Primer Set, from mouse