Merck
HER2 Overexpression in Retinoblastoma: A Potential Therapeutic Target?
Ocular oncology and pathology (2017-12-13)
David Cordeiro Sousa, Pablo Zoroquiain, Maria Eugenia Orellana, Ana Beatriz Dias, Evangelina Esposito, Miguel N Burnier
RESUMEN
Retinoblastoma (RB) is the most common primary intraocular malignancy. Current therapies are associated with high morbidity in the short- and long-term. Human epidermal growth factor receptor 2 (HER2) is a transmembrane protein detected in 15-30% of breast cancers, but it has also been described in other malignancies. Recently, it has been claimed that a truncated version of this protein is expressed in RB, responsive to directed therapies in vitro. We scored HER2 overexpression in RB tissue samples and discussed its potential clinical utility. HER2 overexpression was investigated using immunohistochemistry; the overexpression was evaluated with a score ranging from 0 to 3+ according to the membranous staining pattern in archival formalin-fixed, paraffin-embedded RBs. A total of 60 RB cases and a RB cell line (Y79) were considered. The mean age at enucleation was 31.6 ± 31.5 months. The mean time from diagnosis to enucleation was 11.8 ± 11.2 months (range 1-44). Five (8%) cases were multifocal. HER2 overexpression was negative in all RB cases (49 cases scored 0 and 11 scored 1+) and in the Y79 cell line. Overall, we were not able to demonstrate the overexpression of HER2. Further studies should clarify and better elucidate the potential role of HER2-targeted therapies in RB.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-ERBB2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution