GC Analysis of Volatiles in Water on Equity-1 after SPME using 30 μm PDMS Fiber
Materials
SPME fiber
SPME Fiber Assembly
30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hubCONDITIONS
sample/matrix
0.7 mL water spiked with 2-600 μg/liter each analyte, 0.25 g NaCl added
SPME fiber
30 μm PDMS (57309)
extraction
headspace, 12 sec (no stirring), ambient temp.
desorption process
2-3 min, 250 °C (0.75-1.0 mm I.D. inj. port liner)
column
Equity-1, 10 m × 0.20 mm I.D., 1.2 μm (available on request)
oven
70 °C (0.2 min) to 180 °C at 50 °C/min
detector
FID, 250 °C
Description
Legal Information
null