Merck
  • Characterization of ectonucleoside triphosphate diphosphohydrolase (E-NTPDase; EC 3.6.1.5) activity in mouse peritoneal cavity cells.
Characterization of ectonucleoside triphosphate diphosphohydrolase (E-NTPDase; EC 3.6.1.5) activity in mouse peritoneal cavity cells.
Cell biochemistry and function (2017-09-06)
Dhébora Albuquerque Dias, Bruna de Barros Penteado, Lucas Derbocio Dos Santos, Pedro Mendes Dos Santos, Carla Cardozo Pinto Arruda, Maria Rosa Chitolina Schetinger, Daniela Bitencourt Rosa Leal, Jeandre Augusto Dos Santos Jaques
ABSTRACT
This study aimed to characterize the activity of ectonucleoside triphosphate diphosphohydrolase (E-NTPDase; EC 3.6.1.5) in peritoneal cavity cells from BALB/c mice. E-NTPDase was activated in the presence of both calcium (1.5mM) and magnesium (1.5mM) ions. However, the activity was higher in the presence of Ca2+ . A pH of 8.5 and temperature of 37°C were the optimum conditions for catalysis. The apparent Km values were 0.51mM and 0.66mM for the hydrolysis of adenosine triphosphate (ATP) and adenosine diphosphate (ADP), respectively. The Vmax values were 136.4 and 120.8 nmol Pi/min/mg of protein for ATPase and ADPase activity, respectively. Nucleotide hydrolysis was inhibited in the presence of sodium azide (20mM, ATP: P < .05; ADP: P < .001), sodium fluoride (20mM; ATP and ADP: P < .001), and suramin (0.3mM; ATP: P < .01; ADP: P < .05), which is a known profile for NTPDase inhibition. Although all of the diphosphate and triphosphate nucleotides that were tested were hydrolyzed, enzyme activity was increased when adenine nucleotides were used as substrates. Finally, we stress that knowledge of the E-NTPDase catalytic biochemical properties in mouse peritoneal cavity cells is indispensable for properly determining its activity, as well as to fully understand the immune response profile in both healthy and sick cells.
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Uridine 5′-(trihydrogen diphosphate) sodium salt from Saccharomyces cerevisiae, 95-100%
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)