Merck
  • In vivo probing of nascent RNA structures reveals principles of cotranscriptional folding.
In vivo probing of nascent RNA structures reveals principles of cotranscriptional folding.
Nucleic acids research (2017-09-22)
Danny Incarnato, Edoardo Morandi, Francesca Anselmi, Lisa M Simon, Giulia Basile, Salvatore Oliviero
ABSTRACT
Defining the in vivo folding pathway of cellular RNAs is essential to understand how they reach their final native conformation. We here introduce a novel method, named Structural Probing of Elongating Transcripts (SPET-seq), that permits single-base resolution analysis of transcription intermediates' secondary structures on a transcriptome-wide scale, enabling base-resolution analysis of the RNA folding events. Our results suggest that cotranscriptional RNA folding in vivo is a mixture of cooperative folding events, in which local RNA secondary structure elements are formed as they get transcribed, and non-cooperative events, in which 5'-halves of long-range helices get sequestered into transient non-native interactions until their 3' counterparts have been transcribed. Together our work provides the first transcriptome-scale overview of RNA cotranscriptional folding in a living organism.
Product Description
Glass beads, acid-washed, 150-212 μm (70-100 U.S. sieve)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)