Merck
  • The novel intracellular protein CREG inhibits hepatic steatosis, obesity, and insulin resistance.
The novel intracellular protein CREG inhibits hepatic steatosis, obesity, and insulin resistance.
Hepatology (Baltimore, Md.) (2017-05-17)
Quan-Yu Zhang, Ling-Ping Zhao, Xiao-Xiang Tian, Cheng-Hui Yan, Yang Li, Yan-Xia Liu, Pi-Xiao Wang, Xiao-Jing Zhang, Ya-Ling Han
ABSTRACT
Cellular repressor of E1A-stimulated genes (CREG), a novel cellular glycoprotein, has been identified as a suppressor of various cardiovascular diseases because of its capacity to reduce hyperplasia, maintain vascular homeostasis, and promote endothelial restoration. However, the effects and mechanism of CREG in metabolic disorder and hepatic steatosis remain unknown. Here, we report that hepatocyte-specific CREG deletion dramatically exacerbates high-fat diet and leptin deficiency-induced (ob/ob) adverse effects such as obesity, hepatic steatosis, and metabolic disorders, whereas a beneficial effect is conferred by CREG overexpression. Additional experiments demonstrated that c-Jun N-terminal kinase 1 (JNK1) but not JNK2 is largely responsible for the protective effect of CREG on the aforementioned pathologies. Notably, JNK1 inhibition strongly prevents the adverse effects of CREG deletion on steatosis and related metabolic disorders. Mechanistically, CREG interacts directly with apoptosis signal-regulating kinase 1 (ASK1) and inhibits its phosphorylation, thereby blocking the downstream MKK4/7-JNK1 signaling pathway and leading to significantly alleviated obesity, insulin resistance, and hepatic steatosis. Importantly, dramatically reduced CREG expression and hyperactivated JNK1 signaling was observed in the livers of nonalcoholic fatty liver disease (NAFLD) patients, suggesting that CREG might be a promising therapeutic target for NAFLD and related metabolic diseases. The results of our study provides evidence that CREG is a robust suppressor of hepatic steatosis and metabolic disorders through its direct interaction with ASK1 and the resultant inactivation of ASK1-JNK1 signaling. This study offers insights into NAFLD pathogenesis and its complicated pathologies, such as obesity and insulin resistance, and paves the way for disease treatment through targeting CREG. (Hepatology 2017;66:834-854).
Sigma-Aldrich
Anti-phospho-IRS1 (Tyr608) mouse/ (Tyr612) human Antibody, Upstate®, from rabbit
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Anti-phospho-p70 S6 Kinase (Thr389) Antibody, clone 10G7.1, clone 10G7.1, from mouse
Sigma-Aldrich
Anti-phospho-mTOR (Ser2159) Antibody, from rabbit, purified by affinity chromatography