Merck
  • Embryos cultured in a time-lapse system result in superior treatment outcomes: a strict matched pair analysis.
Embryos cultured in a time-lapse system result in superior treatment outcomes: a strict matched pair analysis.
Human fertility (Cambridge, England) (2016-11-26)
Amy Barrie, Roy Homburg, Garry McDowell, Jeremy Brown, Charles Kingsland, Stephen Troup
ABSTRACT
A retrospective strict matched-pair analysis of 728 treatment cycles between January 2011 and September 2014 was performed. A total of 364 treatment cycles, where all embryos were cultured and examined in EmbryoScope
Sigma-Aldrich
Cetrorelix acetate, ≥98% (HPLC)