Merck
  • A novel LCMSMS method for quantitative measurement of short-chain fatty acids in human stool derivatized with
A novel LCMSMS method for quantitative measurement of short-chain fatty acids in human stool derivatized with
Journal of pharmaceutical and biomedical analysis (2017-02-09)
James Chun Yip Chan, Dorinda Yan Qin Kioh, Gaik Chin Yap, Bee Wah Lee, Eric Chun Yong Chan
ABSTRACT
A novel liquid chromatography tandem mass spectrometry (LCMSMS) method for the quantitative measurement of gut microbial-derived short-chain fatty acids (SCFAs) in human infant stool has been developed and validated. Baseline chromatographic resolution was achieved for 12 SCFAs (acetic, butyric, caproic, 2,2-dimethylbutyric, 2-ethylbutyric, isobutyric, isovaleric, 2-methylbutyric, 4-methylvaleric, propionic, pivalic and valeric acids) within an analysis time of 15min. A novel sequential derivatization of endogenous and spiked SCFAs in stool via
Sigma-Aldrich
2,2-Dimethylbutyric acid, 96%
Sigma-Aldrich
3,3-Dimethylbutyric acid, 98%
Sigma-Aldrich
Pivalic acid, 99%