Merck
  • Regulation of Chemokine Signal Integration by Activator of G-Protein Signaling 4 (AGS4).
Regulation of Chemokine Signal Integration by Activator of G-Protein Signaling 4 (AGS4).
The Journal of pharmacology and experimental therapeutics (2017-01-08)
William G Robichaux, Melissa Branham-O'Connor, Il-Young Hwang, Ali Vural, Johne H Kehrl, Joe B Blumer
ABSTRACT
Activator of G-protein signaling 4 (AGS4)/G-protein signaling modulator 3 (Gpsm3) contains three G-protein regulatory (GPR) motifs, each of which can bind G
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Millipore
Thioglycollate Broth (USP Alternative), suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture