Merck
Compound-Specific Sulfur Isotope Analysis of Petroleum Gases.
Analytical chemistry (2017-02-15)
Ward Said-Ahmad, Kenneth Wong, Monaca Mcnall, Lubna Shawar, Tracey Jacksier, Courtney Turich, Artur Stankiewicz, Alon Amrani
ABSTRACT
We describe a simple, sensitive, and robust method for sulfur isotope ratio (
Sigma-Aldrich
Carbon disulfide, anhydrous, ≥99%
Sigma-Aldrich
Ethyl methyl sulfide, 96%
Sigma-Aldrich
Thiophene, ≥99%