Merck
  • Linkage of beta1-adrenergic stimulation to apoptotic heart cell death through protein kinase A-independent activation of Ca2+/calmodulin kinase II.
Linkage of beta1-adrenergic stimulation to apoptotic heart cell death through protein kinase A-independent activation of Ca2+/calmodulin kinase II.
The Journal of clinical investigation (2003-03-06)
Wei-Zhong Zhu, Shi-Qiang Wang, Khalid Chakir, Dongmei Yang, Tong Zhang, Joan Heller Brown, Eric Devic, Brian K Kobilka, Heping Cheng, Rui-Ping Xiao
ABSTRACT
beta(1)-adrenergic receptor (beta(1)AR) stimulation activates the classic cAMP/protein kinase A (PKA) pathway to regulate vital cellular processes from the change of gene expression to the control of metabolism, muscle contraction, and cell apoptosis. Here we show that sustained beta(1)AR stimulation promotes cardiac myocyte apoptosis by activation of Ca(2+)/calmodulin kinase II (CaMKII), independently of PKA signaling. beta(1)AR-induced apoptosis is resistant to inhibition of PKA by a specific peptide inhibitor, PKI14-22, or an inactive cAMP analogue, Rp-8-CPT-cAMPS. In contrast, the beta(1)AR proapoptotic effect is associated with non-PKA-dependent increases in intracellular Ca(2+) and CaMKII activity. Blocking the L-type Ca(2+) channel, buffering intracellular Ca(2+), or inhibiting CaMKII activity fully protects cardiac myocytes against beta(1)AR-induced apoptosis, and overexpressing a cardiac CaMKII isoform, CaMKII-deltaC, markedly exaggerates the beta(1)AR apoptotic effect. These findings indicate that CaMKII constitutes a novel PKA-independent linkage of beta(1)AR stimulation to cardiomyocyte apoptosis that has been implicated in the overall process of chronic heart failure.
Sigma-Aldrich
CAMK2δ, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution