Merck
Matrix metalloproteinases and the regulation of tissue remodelling.
Nature reviews. Molecular cell biology (2007-02-24)
Andrea Page-McCaw, Andrew J Ewald, Zena Werb
ABSTRACT
Matrix metalloproteinases (MMPs) were discovered because of their role in amphibian metamorphosis, yet they have attracted more attention because of their roles in disease. Despite intensive scrutiny in vitro, in cell culture and in animal models, the normal physiological roles of these extracellular proteases have been elusive. Recent studies in mice and flies point to essential roles of MMPs as mediators of change and physical adaptation in tissues, whether developmentally regulated, environmentally induced or disease associated.
Sigma-Aldrich
MMP-2 pre-activated human, recombinant, ≥1,000 pmol/min/μg, expressed in HEK 293 cells
Sigma-Aldrich
MMP-2 human, recombinant, ≥1,000 pmol/min/μg, expressed in HEK 293 cells