Merck
  • Assessing the evolutionary history of the class Synurophyceae (Heterokonta) using molecular, morphometric, and paleobiological approaches.
Assessing the evolutionary history of the class Synurophyceae (Heterokonta) using molecular, morphometric, and paleobiological approaches.
American journal of botany (2015-06-24)
Peter A Siver, Bok Yeon Jo, Jong Im Kim, Woongghi Shin, Anne Marie Lott, Alexander P Wolfe
ABSTRACT
Heterokont algae of the class Synurophyceae, characterized by distinctive siliceous scales that cover the surface of the cell, are ecologically important in inland waters, yet their evolutionary history remains enigmatic. We explore phylogenetic relationships within this group of algae relative to geologic time, with a focus on evolution of siliceous components. We combined an expansive five-gene and time-calibrated molecular phylogeny of synurophyte algae with an extensive array of fossil specimens from the middle Eocene to infer evolutionary trends within the group. The group originated in the Jurassic approximately 157 million years ago (Ma), with the keystone genera Mallomonas and Synura diverging during the Early Cretaceous at 130 Ma. Mallomonas further splits into two major subclades, signaling the evolution of the V-rib believed to aid in the spacing and organization of scales on the cell covering. Synura also diverges into two primary subclades, separating taxa with forward-projecting spines on the scale from those with a keel positioned on the scale proper. Approximately one third of the fossil species are extinct, whereas the remaining taxa are linked to modern congeners. The taxonomy of synurophytes, which relies extensively on the morphology of the siliceous components, is largely congruent with molecular analyses. Scales of extinct synurophytes were significantly larger than those of modern taxa and may have played a role in their demise. In contrast, many fossil species linked to modern lineages were smaller in the middle Eocene, possibly reflecting growth in the greenhouse climatic state that characterized this geologic interval.
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Potassium dichromate solution, 1/60 M