Merck
  • Adseverin plays a role in osteoclast differentiation and periodontal disease-mediated bone loss.
Adseverin plays a role in osteoclast differentiation and periodontal disease-mediated bone loss.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-02-15)
Hongwei Jiang, Yongqiang Wang, Ana Viniegra, Corneliu Sima, Christopher A McCulloch, Michael Glogauer
ABSTRACT
Osteoclast differentiation and function are highly dependent on the assembly and turnover of actin filaments, but little is known about the roles of actin binding proteins in these processes. Adseverin (Ads), a member of the gelsolin superfamily of actin capping and severing proteins, regulates actin filament turnover and can regulate the turnover of cortical actin filaments of chromaffin cells during exocytosis. Using a conditional Ads knockout mouse model, we confirmed our previous finding in cultured cells that Ads plays a role in osteoclastogenesis (OCG) and actin cytoskeletal organization in osteoclasts. Here we show that Ads is required for osteoclast formation and that when alveolar bone resorption is experimentally induced in mice, genetic deletion of Ads prevents osteoclast-mediated bone loss. Further, when Ads-null osteoclasts are cultured, they exhibit defective OCG, disorganized podosome-based actin filament superstructures, and decreased bone resorption. Reintroduction of Ads into Ads-null osteoclast precursor cells restored these osteoclast defects. Collectively, these data demonstrate a unique and osteoclast-specific role for Ads in OCG and osteoclast function.
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O