Merck
  • Kinetics and energy efficiency for the degradation of 1,4-dioxane by electro-peroxone process.
Kinetics and energy efficiency for the degradation of 1,4-dioxane by electro-peroxone process.
Journal of hazardous materials (2015-04-12)
Huijiao Wang, Belal Bakheet, Shi Yuan, Xiang Li, Gang Yu, Seiichi Murayama, Yujue Wang
ABSTRACT
Degradation of 1,4-dioxane by ozonation, electrolysis, and their combined electro-peroxone (E-peroxone) process was investigated. The E-peroxone process used a carbon-polytetrafluorethylene cathode to electrocatalytically convert O2 in the sparged ozone generator effluent (O2 and O3 gas mixture) to H2O2. The electro-generated H2O2 then react with sparged O3 to yield aqueous OH, which can in turn oxidize pollutants rapidly in the bulk solution. Using p-chlorobenzoic acid as OH probe, the pseudo-steady concentration of OH was determined to be ∼0.744×10(-9)mM in the E-peroxone process, which is approximately 10 and 186 times of that in ozonation and electrolysis using a Pt anode. Thanks to its higher OH concentration, the E-peroxone process eliminated 96.6% total organic carbon (TOC) from a 1,4-dioxane solution after 2h treatment with a specific energy consumption (SEC) of 0.376kWhg(-1) TOCremoved. In comparison, ozonation and electrolysis using a boron-doped diamond anode removed only ∼6.1% and 26.9% TOC with SEC of 2.43 and 0.558kWhg(-1) TOCremoved, respectively. The results indicate that the E-peroxone process can significantly improve the kinetics and energy efficiency for 1,4-dioxane mineralization as compared to the two individual processes. The E-peroxone process may thus offer a highly effective and energy-efficient alternative to treat 1,4-dioxane wastewater.
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC
Sigma-Aldrich
1,4-Dioxane, SAJ first grade
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, JIS special grade, ≥99.0%