Merck
  • Aquaporin 3 knockdown suppresses tumour growth and angiogenesis in experimental non-small cell lung cancer.
Aquaporin 3 knockdown suppresses tumour growth and angiogenesis in experimental non-small cell lung cancer.
Experimental physiology (2014-05-08)
Hui Xia, Yong-Fu Ma, Chang-Hai Yu, Ying-Jie Li, Jian Tang, Jing-Bo Li, Ying-Nan Zhao, Yang Liu
ABSTRACT
Non-small cell lung cancer (NSCLC) is one of the most common diseases encountered in medical oncology practice. The aim of the present study was to test the antitumour effects of short-hairpin RNA targeting aquaporin 3 (AQP3) in experimental NSCLC. Expression of AQP3 was suppressed in human A549 and H1299 NSCLC cell lines by short-hairpin RNA-mediated silencing. Therapeutic effects were assessed by examining tumorigenicity using a subcutaneous xenograft mouse model of NSCLC. Aquaporin 3 knockdown inhibited tumour growth and prolonged survival of mice with tumours. Aquaporin 3 knockdown suppressed tumour proliferation, marked by enhanced expression of p53, an increased ratio of cleaved caspase 3 to pro-caspase 3 and reduced expression of proliferating cell nuclear antigen and B-cell lymphoma-2 (bcl-2). Aquaporin 3 knockdown inhibited tumour angiogenesis, marked by decreased CD31 immunostaining and reduced expression of hypoxia-inducible factor-2α and vascular endothelial growth factor. Aquaporin 3 knockdown reduced cellular glycerol content and suppressed mitochondrial ATP formation. Aquaporin 3 knockdown in vitro significantly suppressed activities of matrix metalloproteinases MMP2 and MMP9, reduced AKT phosphorylation and decreased cell invasiveness of A549 and H1299 cells. In conclusion, AQP3 knockdown suppressed tumour growth and reduced angiogenesis in human NSCLS xenografts. Aquaporin 3 could thus be envisaged as a novel therapeutic target for NSCLC.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting human AQP3
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Aqp3
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Supelco
Glycerol, analytical standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%