Merck
  • A small-molecule inhibitor of Haspin alters the kinetochore functions of Aurora B.
A small-molecule inhibitor of Haspin alters the kinetochore functions of Aurora B.
The Journal of cell biology (2012-10-17)
Anna De Antoni, Stefano Maffini, Stefan Knapp, Andrea Musacchio, Stefano Santaguida
ABSTRACT
By phosphorylating Thr3 of histone H3, Haspin promotes centromeric recruitment of the chromosome passenger complex (CPC) during mitosis. Aurora B kinase, a CPC subunit, sustains chromosome bi-orientation and the spindle assembly checkpoint (SAC). Here, we characterize the small molecule 5-iodotubercidin (5-ITu) as a potent Haspin inhibitor. In vitro, 5-ITu potently inhibited Haspin but not Aurora B. Consistently, 5-ITu counteracted the centromeric localization of the CPC without affecting the bulk of Aurora B activity in HeLa cells. Mislocalization of Aurora B correlated with dephosphorylation of CENP-A and Hec1 and SAC override at high nocodazole concentrations. 5-ITu also impaired kinetochore recruitment of Bub1 and BubR1 kinases, and this effect was reversed by concomitant inhibition of phosphatase activity. Forcing localization of Aurora B to centromeres in 5-ITu also restored Bub1 and BubR1 localization but failed to rescue the SAC override. This result suggests that a target of 5-ITu, possibly Haspin itself, may further contribute to SAC signaling downstream of Aurora B.
Sigma-Aldrich
5-Iodotubercidin, ≥85%, solid
Sigma-Aldrich
Tubercidin, from Streptomyces tubercidicus, ~95%
Sigma-Aldrich
Anti-phospho-CENP-A (Ser7) Antibody, clone NL41, rabbit monoclonal, culture supernatant, clone NL41, Upstate®
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit