Merck
  • Oil spills abatement: factors affecting oil uptake by cellulosic fibers.
Oil spills abatement: factors affecting oil uptake by cellulosic fibers.
Environmental science & technology (2012-06-26)
Katharine C Payne, Colby D Jackson, Carlos E Aizpurua, Orlando J Rojas, Martin A Hubbe
ABSTRACT
Wood-derived cellulosic fibers prepared in different ways were successfully employed to absorb simulated crude oil, demonstrating their possible use as absorbents in the case of oil spills. When dry fibers were used, the highest sorption capacity (six parts of oil per unit mass of fiber) was shown by bleached softwood kraft fibers, compared to hardwood bleached kraft and softwood chemithermomechanical pulp(CTMP) fibers. Increased refining of CTMP fibers decreased their oil uptake capacity. When the fibers were soaked in water before exposure to the oil, the ability of the unmodified kraft fibers to sorb oil was markedly reduced, whereas the wet CTMP fibers were generally more effective than the wet kraft fibers. Predeposition of lignin onto the surfaces of the bleached kraft fibers improved their ability to take up oil when wet. Superior ability to sorb oil in the wet state was achieved by pretreating the kraft fibers with a hydrophobic sizing agent, alkenylsuccinic anhydride (ASA). Contact angle tests on a model cellulose surface showed that some of the sorption results onto wetted fibers could be attributed to the more hydrophobic nature of the fibers after treatment with either lignin or ASA.
Sigma-Aldrich
Succinic anhydride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)