Merck
  • Genistein ameliorates adverse cardiac effects induced by arsenic trioxide through preventing cardiomyocytes apoptosis.
Genistein ameliorates adverse cardiac effects induced by arsenic trioxide through preventing cardiomyocytes apoptosis.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2013-02-01)
Yuhua Fan, Chen Wang, Yong Zhang, Pengzhou Hang, Yan Liu, Zhenwei Pan, Ning Wang, Zhimin Du
ABSTRACT
Arsenic trioxide (As2O3) is a highly effective agent for treatment of acute promyelocytic leukemia (APL). However, consecutively administered As2O3 induces serious adverse cardiac effects, including long QT syndrome (LQTs) and even sudden cardiac death. Previous studies have shown that genistein (Gen) exerts anti-oxidant, anti-inflammatory, and anti-apoptotic effects. The present study aimed to explore the potential protective effects of Gen on As2O3-induced adverse cardiac effects, and to elucidate the underlying molecular mechanisms. A rat model of As2O3-induced QT prolongation was generated by intravenous injection with As2O3. Surface electrocardiogram (ECG) and hemodynamics were employed to assess the LQTs and cardiac function. Intracellular calcium concentration ([Ca(2+)]i) and mitochondrial membrane potential (Δψm) were measured by confocal microscopy, and cardiomyocytes apoptosis were assessed by TUNEL assay. Western blot was applied to determine protein levels. We found for the first time that treatment with Gen significantly reversed LQTs and dose-dependently improved As2O3-induced impairment of cardiac function. As2O3 elevated [Ca(2+)]i and Gen mitigated this effect. Meanwhile, Gen significantly reversed As2O3-mediated cardiomyocytes apoptosis. Furthermore, Gen dose-dependently inhibited the phosphorylated JNK and p38-MAPK (pp38-MAPK), and blocked Δψm collapse, and further decreased cleaved caspase-3. Gen protects against the adverse cardiac effects of As2O3 partly by mitigating cardiomyocytes apoptosis induced by As2O3 through attenuating intracellular calcium overload and downregulating protein expression of p-JNK and pp38-MAPK to ameliorate the damage of Δψm leading to suppression of caspase-3 activation. Gen might be used as an adjunction therapy in APL patients receiving As2O3 treatment to avoid, at least to minimize, the adverse cardiac effects of As2O3.
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Sigma-Aldrich
Arsenic(III) oxide, SAJ first grade, ≥99.0%
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis