Merck
The influence of hydroquinone on tyrosinase kinetics.
Bioorganic & medicinal chemistry (2012-06-16)
Michael R L Stratford, Christopher A Ramsden, Patrick A Riley
ABSTRACT
In vitro studies, using combined spectrophotometry and oximetry together with hplc/ms examination of the products of tyrosinase action demonstrate that hydroquinone is not a primary substrate for the enzyme but is vicariously oxidised by a redox exchange mechanism in the presence of either catechol, L-3,4-dihydroxyphenylalanine or 4-ethylphenol. Secondary addition products formed in the presence of hydroquinone are shown to stimulate, rather than inhibit, the kinetics of substrate oxidation.
Sigma-Aldrich
4-Ethylphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
Pricing and availability is not currently available.