Merck
  • Growth, photosynthesis and antioxidant responses of endophyte infected and non-infected rice under lead stress conditions.
Growth, photosynthesis and antioxidant responses of endophyte infected and non-infected rice under lead stress conditions.
Journal of hazardous materials (2012-02-24)
Xuemei Li, Ning Bu, Yueying Li, Lianju Ma, Shigang Xin, Lihong Zhang
ABSTRACT
An endophytic fungus was tested in rice (Oryza sativa L.) exposed to four levels of lead (Pb) stress (0, 50, 100 and 200 μM) to assess effects on plant growth, photosynthesis and antioxidant enzyme activity. Under Pb stress conditions, endophyte-infected seedlings had greater shoot length but lower root length compared to non-infected controls, and endophyte-infected seedlings had greater dry weight in the 50 and 100 μM Pb treatments. Under Pb stress conditions, chlorophyll and carotenoid levels were significantly higher in the endophyte-infected seedlings. Net photosynthetic rate, transpiration rate and water use efficiency were significantly higher in endophyte-infected seedlings in the 50 and 100 μM Pb treatments. In addition, chlorophyll fluorescence parameters Fv/Fm and Fv/Fo were higher in the infected seedlings compared to the non-infected seedlings under Pb stress. Malondialdehyde accumulation was induced by Pb stress, and it was present in higher concentration in non-infected seedlings under higher concentrations of Pb (100 and 200 μM). Antioxidant activity was either higher or unchanged in the infected seedlings due to responses to the different Pb concentrations. These results suggest that the endophytic fungus improved rice growth under moderate Pb levels by enhancing photosynthesis and antioxidant activity relative to non-infected rice.
Sigma-Aldrich
Lead(II) nitrate, ≥99.95% trace metals basis
Sigma-Aldrich
Lead standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Pb, 1000 ppm Pb
Sigma-Aldrich
Lead(II) nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Lead(II) nitrate, 99.999% trace metals basis