Merck
  • Study of Trypanosoma cruzi epimastigote cell death by NMR-visible mobile lipid analysis.
Study of Trypanosoma cruzi epimastigote cell death by NMR-visible mobile lipid analysis.
Parasitology (2012-01-06)
Diego Benitez, Horacio Pezaroglo, Verónica Martínez, Gabriela Casanova, Gonzalo Cabrera, Norbel Galanti, Mercedes González, Hugo Cerecetto
ABSTRACT
Cell death mechanisms in Trypanosoma cruzi have not been disclosed in detail though different conventional techniques have been used in the classification of parasite-cell death type. Nuclear magnetic resonance (NMR) has successfully been used as a tool to evaluate the onset of apoptosis in a number of higher eukaryote-cell models analysing the ratio of CH(2)/CH(3) integration from the visible mobile lipids (VML). Surprisingly, this versatile non-invasive spectroscopy technique has never been employed with this purpose in T. cruzi. In the present study it is shown that under different parasite death-conditions the ratio CH(2)/CH(3) varied drastically. Thus, T. cruzi epimastigotes in apoptotic conditions increase significantly this ratio while in necrotic as well as in autophagic situations the parasites maintain the VML, CH(2)/CH(3) ratio, in normal values. Additionally, other VML markers commonly used in these studies, such as the change in the region of methyl-choline moiety, -N(+)(CH(3))(3), exhibited different particular patterns according to the type of cell death. Our results suggest that the (1)H NMR-VML technique is an adequate tool to discriminate different T. cruzi death pathways.
Sigma-Aldrich
Nifurtimox, ≥98% (HPLC)