Merck
  • Role of metabolism by the human intestinal microflora in arbutin-induced cytotoxicity in HepG2 cell cultures.
Role of metabolism by the human intestinal microflora in arbutin-induced cytotoxicity in HepG2 cell cultures.
Biochemical and biophysical research communications (2011-09-06)
Tilak Khanal, Hyung Gyun Kim, Yong Pil Hwang, Min Jeong Kong, Mi Jeong Kang, Hee Kyung Yeo, Dong Hyun Kim, Tae Cheon Jeong, Hye Gwang Jeong
ABSTRACT
A possible role for metabolism by the human intestinal microflora in arbutin-induced cytotoxicity was investigated using human hepatoma HepG2 cells. When the cytotoxic effects of arbutin and hydroquinone (HQ), a deglycosylated metabolite of arbutin, were compared, HQ was more toxic than arbutin. Incubation of arbutin with a human fecal preparation could produce HQ. Following incubation of arbutin with a human fecal preparation for metabolic activation, the reaction mixture was filter-sterilized to test its toxic effects on HepG2 cells. The mixture induced cytotoxicity in HepG2 cells in a concentration-dependent manner. In addition, the mixture considerably inhibited expression of Bcl-2 together with an increase in Bax expression. Likewise, activation stimulated cleavage of caspase-3 and production of reactive oxygen species in HepG2 cell cultures. Furthermore, induction of apoptosis by the intestinal microflora reaction mixture was confirmed by the terminal deoxynucleotidyltransferase-mediated dUTP-biotin nick-end labeling assay. Taken together, these findings suggest that the human intestinal microflora is capable of metabolizing arbutin to HQ, which can induce apoptosis in mammalian cells.
Product Description
Arbutin, European Pharmacopoeia (EP) Reference Standard
Supelco
Arbutin, analytical standard
Sigma-Aldrich
Arbutin, ≥98% (HPLC)