Merck
Penicillamine-induced pemphigus erythematosus.
International journal of dermatology (1990-04-01)
M J Willemsen, A L De Coninck, L E De Raeve, D I Roseeuw
ABSTRACT
Pemphigus erythematosus occurred in a patient with rheumatoid arthritis who was treated with D-penicillamine. The skin lesions appeared 4 months after the onset of D-penicillamine treatment and persisted 14 years after cessation of this drug. Topical betamethasone dipropionate applications resulted in complete regression of the cutaneous lesions.
Sigma-Aldrich
D-Penicillamine disulfide, 97%