Merck
  • Chiral anion-mediated asymmetric ring opening of meso-aziridinium and episulfonium ions.
Chiral anion-mediated asymmetric ring opening of meso-aziridinium and episulfonium ions.
Journal of the American Chemical Society (2008-10-22)
Gregory L Hamilton, Toshio Kanai, F Dean Toste
ABSTRACT
Reactions proceeding through cationic intermediates that lack a Lewis or Brønsted basic site present a challenge for traditional asymmetric catalysis based on chiral metals or organocatalysts. We present an enantioselective ring opening of tetrasubstituted meso-aziridinium ions with alcohol nucleophiles proceeding through a chiral ion pair with a binaphthol-phosphate anion. The reaction is initiated by silver-induced ring closure of beta-chloroamines using the Ag salt of the chiral anion as in situ generated catalyst. Use of insoluble Ag2CO3 as silver source is essential to obtain high enantioselectivity; we believe the chiral phosphate acts as a "chiral anion phase transfer catalyst" to bring silver ion into the organic phase. The chiral anion concept can also be extended to the related asymmetric opening of meso-episulfonium ions generated by protonation of trichloroacetimidates vicinal to sulfides.
Sigma-Aldrich
Silver carbonate, 99%
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)