Merck
  • Maximising opportunities in supercritical chemistry: the continuous conversion of levulinic acid to gamma-valerolactone in CO(2).
Maximising opportunities in supercritical chemistry: the continuous conversion of levulinic acid to gamma-valerolactone in CO(2).
Chemical communications (Cambridge, England) (2007-11-09)
Richard A Bourne, James G Stevens, Jie Ke, Martyn Poliakoff
ABSTRACT
Phase behaviour is manipulated during the hydrogenation of aqueous levulinic acid in supercritical CO(2) to separate almost pure gamma-valerolactone from water and unreacted acid with reduced energy requirements compared to conventional processing.
Sigma-Aldrich
γ-Valerolactone, natural, ≥95%, FG
Sigma-Aldrich
γ-Valerolactone, ≥99%, FCC, FG
Sigma-Aldrich
γ-Valerolactone, ReagentPlus®, 99%