Merck
  • Lithium diisopropylamide solvated by hexamethylphosphoramide: substrate-dependent mechanisms for dehydrobrominations.
Lithium diisopropylamide solvated by hexamethylphosphoramide: substrate-dependent mechanisms for dehydrobrominations.
Journal of the American Chemical Society (2006-11-30)
Yun Ma, Antonio Ramirez, Kanwal Jit Singh, Ivan Keresztes, David B Collum
ABSTRACT
Lithium diisopropylamide-mediated dehydrobrominations of exo-2-bromonorbornane, 1-bromocyclooctene, and cis-4-bromo-tert-butylcyclohexane were studied in THF solutions and THF solutions with added hexamethylphosphoramide (HMPA). Rate studies reveal a diverse array of mechanisms based on mono-, di-, and trisolvated monomers as well as triple ions. The results are contrasted with analogous eliminations in THF in the absence of HMPA.
Sigma-Aldrich
Hexamethylphosphoramide, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexamethylphosphoramide, 99%
Sigma-Aldrich
Hexamethylphosphoramide, absolute, over molecular sieve (H2O ≤0.03%), ≥98.0% (GC)
Sigma-Aldrich
Diisopropylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Supelco
Diisopropylamine, analytical standard
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)