Merck
  • Biochemical evaluation of an infant with hypoglycemia resulting from a novel de novo mutation of the GLUD1 gene and hyperinsulinism-hyperammonemia syndrome.
Biochemical evaluation of an infant with hypoglycemia resulting from a novel de novo mutation of the GLUD1 gene and hyperinsulinism-hyperammonemia syndrome.
Journal of pediatric endocrinology & metabolism : JPEM (2011-09-22)
Shanti Balasubramaniam, Ritika Kapoor, Janet Hua Hong Yeow, Poi Geok Lim, Sarah Flanagan, Sian Ellard, Khalid Hussain
ABSTRACT
Hyperinsulinism-hyperammonemia syndrome (HI/HA) (OMIM 606762), the second most common form of congenital hyperinsulinism (CHI) is associated with activating missense mutations in the GLUD1 gene, which encodes the mitochondrial matrix enzyme, glutamate dehydrogenase (GDH). Patients present with recurrent symptomatic postprandial hypoglycemia following protein-rich meals (leucine-sensitive hypoglycemia) as well as fasting hypoglycemia accompanied by asymptomatic elevations of plasma ammonia. In contrast to other forms of CHI, the phenotype is reported to be milder thus escaping recognition for the first few months of life. Early diagnosis and appropriate management are essential to avoid the neurodevelopmental consequences including epilepsy and learning disabilities which are prevalent in this disorder. We report an infant presenting with afebrile seizures secondary to hyperinsulinemic hypoglycemia resulting from a novel de novo mutation of the GLUD1 gene.
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type III, lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type I, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
L-Glutamic Dehydrogenase from bovine liver, Type II, 50% glycerol solution, ≥35 units/mg protein