Merck
  • PLIC proteins or ubiquilins regulate autophagy-dependent cell survival during nutrient starvation.
PLIC proteins or ubiquilins regulate autophagy-dependent cell survival during nutrient starvation.
EMBO reports (2009-01-17)
Elsa-Noah N'Diaye, Kimberly K Kajihara, Ivy Hsieh, Hiroshi Morisaki, Jayanta Debnath, Eric J Brown
ABSTRACT
Ubiquilins (UBQLNs) are adaptor proteins thought to deliver ubiquitinated substrates to proteasomes. Here, we show a role for UBQLN in autophagy: enforced expression of UBQLN protects cells from starvation-induced death, whereas depletion of UBQLN renders cells more susceptible. The UBQLN protective effect requires the autophagy-related genes ATG5 and ATG7, two essential components of autophagy. The ubiquitin-associated domain of UBQLN mediates both its association with autophagosomes and its protective effect against starvation. Depletion of UBQLN delays the delivery of autophagosomes to lysosomes. This study identifies a new role for UBQLN in regulating the maturation of autophagy, expanding the involvement of ubiquitin-related proteins in this process.
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP