Merck
  • Acute Influenza Infection Promotes Lung Tumor Growth by Reprogramming the Tumor Microenvironment.
Acute Influenza Infection Promotes Lung Tumor Growth by Reprogramming the Tumor Microenvironment.
Cancer immunology research (2023-03-09)
Irati Garmendia, Aditi Varthaman, Solenne Marmier, Mahmud Angrini, Ingrid Matchoua, Aurelie Darbois-Delahousse, Nathalie Josseaume, Pierre-Emmanuel Foy, Lubka T Roumenina, Naïra Naouar, Maxime Meylan, Sophie Sibéril, Jules Russick, Pierre-Emmanuel Joubert, Karen Leroy, Diane Damotte, Audrey Mansuet-Lupo, Marie Wislez, Marco Alifano, Laurie Menger, Ignacio Garcia-Verdugo, Jean-Michel Sallenave, Olivier Lantz, Florent Petitprez, Isabelle Cremer
ABSTRACT
One billion people worldwide get flu every year, including patients with non-small cell lung cancer (NSCLC). However, the impact of acute influenza A virus (IAV) infection on the composition of the tumor microenvironment (TME) and the clinical outcome of patients with NSCLC is largely unknown. We set out to understand how IAV load impacts cancer growth and modifies cellular and molecular players in the TME. Herein, we report that IAV can infect both tumor and immune cells, resulting in a long-term protumoral effect in tumor-bearing mice. Mechanistically, IAV impaired tumor-specific T-cell responses, led to the exhaustion of memory CD8+ T cells and induced PD-L1 expression on tumor cells. IAV infection modulated the transcriptomic profile of the TME, fine-tuning it toward immunosuppression, carcinogenesis, and lipid and drug metabolism. Consistent with these data, the transcriptional module induced by IAV infection in tumor cells in tumor-bearing mice was also found in human patients with lung adenocarcinoma and correlated with poor overall survival. In conclusion, we found that IAV infection worsened lung tumor progression by reprogramming the TME toward a more aggressive state.
Sigma-Aldrich
IgM, Mouse, Mouse IgM Polyclonal Antibody control validated for use in ELISA.
Sigma-Aldrich
Goat Anti-Mouse IgM µ chain Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from goat
Sigma-Aldrich
IgG, Mouse