Merck
  • Lithium ameliorates Niemann-Pick C1 disease phenotypes by impeding STING/SREBP2 activation.
Lithium ameliorates Niemann-Pick C1 disease phenotypes by impeding STING/SREBP2 activation.
iScience (2023-05-02)
Shiqian Han, Qijun Wang, Yongfeng Song, Mao Pang, Chunguang Ren, Jing Wang, Dongwei Guan, Wei Xu, Fangyong Li, Fengchao Wang, Xinyuan Zhou, Carlos Fernández-Hernando, Huiwen Zhang, Dianqing Wu, Zhijia Ye
ABSTRACT
Niemann-Pick disease type C (NP-C) is a genetic lysosomal disorder associated with progressive neurodegenerative phenotypes. Its therapeutic options are very limited. Here, we show that lithium treatment improves ataxia and feeding phenotypes, attenuates cerebellar inflammation and degeneration, and extends survival in Npc1 mouse models. In addition, lithium suppresses STING activation, SREBP2 processing to its mature form and the expression of the target genes in the Npc1 mice and in Npc1-deficient fibroblasts. Lithium impedes STING/SREBP2 transport from the ER to the Golgi, a step required for STING activation and SREBP2 processing, probably by lowering cytosolic calcium concentrations. This effect of lithium on STING/SREBP2 transport provides a mechanistic explanation for lithium's effects on Npc1 mice. Thus, this study reveals a potential therapeutic option for NP-C patients as well as a strategy to reduce active STING/SREBP2 pathway.
Sigma-Aldrich
ALLN, Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-SREBP-2 Antibody, clone 22D5, clone 22D5, from rabbit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse