Merck
  • D4F alleviates the C/EBP homologous protein-mediated apoptosis in glycated high-density lipoprotein-treated macrophages by facilitating autophagy.
D4F alleviates the C/EBP homologous protein-mediated apoptosis in glycated high-density lipoprotein-treated macrophages by facilitating autophagy.
Experimental biology and medicine (Maywood, N.J.) (2021-09-17)
Hua Tian, Zhaoqiang Zhang, Xiaoyan Han, Tianqi Pan, Geru Tao, Peng Jiao, Lei Zhai, Libo Yang, Xiaoxu Wang, Yilin Yao, Shucun Qin, Shutong Yao
ABSTRACT
The present study aimed to investigate the role of D4F, an apolipoprotein A-I mimetic peptide, in macrophage apoptosis induced by the glycated high-density lipoprotein (gly-HDL)-induced endoplasmic reticulum (ER) stress C/EBP homologous protein (CHOP) pathway, and unravel the regulatory role of autophagy in this process. Our results revealed that except for suppressing the accumulation of lipids within RAW264.7 macrophages caused by gly-HDL, D4F inhibited gly-HDL-induced decrease in the cell viability and increase in lactate dehydrogenase leakage and cell apoptosis, which were similar to 4-phenylbutyric acid (PBA, an ER stress inhibitor). Besides, similar to PBA, D4F inhibited gly-HDL-induced ER stress response activation evaluated through the decreased PERK and eIF2α phosphorylation, together with reduced ATF6 nuclear translocation as well as the downregulation of GRP78 and CHOP. Interestingly, D4F facilitated gly-HDL-triggered activation of autophagy, measured as elevated levels of beclin-1, LC3-II, and ATG5 expressions in macrophages. Furthermore, the inhibition effect of D4F on gly-HDL-induced ER stress-CHOP-induced apoptosis of macrophages was restrained after beclin-1 siRNA and 3-methyladenine (3-MA, an inhibitor of autophagy) treatments, while this effect was further reinforced after rapamycin (Rapa, an inducer of autophagy) treatment. Furthermore, administering D4F or Rapa to T2DM mice upregulated LC3-II and attenuated CHOP expression, cell apoptosis, and atherosclerotic lesions. However, the opposite results were obtained when 3-MA was administered to these mice. These results support that D4F effectively protects macrophages against gly-HDL-induced ER stress-CHOP-mediated apoptosis by promoting autophagy.
Sigma-Aldrich
Anti-ATG5 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Beclin-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution