Merck
  • Histone H4 lysine 16 acetylation controls central carbon metabolism and diet-induced obesity in mice.
Histone H4 lysine 16 acetylation controls central carbon metabolism and diet-induced obesity in mice.
Nature communications (2021-10-29)
Cecilia Pessoa Rodrigues, Aindrila Chatterjee, Meike Wiese, Thomas Stehle, Witold Szymanski, Maria Shvedunova, Asifa Akhtar
ABSTRACT
Noncommunicable diseases (NCDs) account for over 70% of deaths world-wide. Previous work has linked NCDs such as type 2 diabetes (T2D) to disruption of chromatin regulators. However, the exact molecular origins of these chronic conditions remain elusive. Here, we identify the H4 lysine 16 acetyltransferase MOF as a critical regulator of central carbon metabolism. High-throughput metabolomics unveil a systemic amino acid and carbohydrate imbalance in Mof deficient mice, manifesting in T2D predisposition. Oral glucose tolerance testing (OGTT) reveals defects in glucose assimilation and insulin secretion in these animals. Furthermore, Mof deficient mice are resistant to diet-induced fat gain due to defects in glucose uptake in adipose tissue. MOF-mediated H4K16ac deposition controls expression of the master regulator of glucose metabolism, Pparg and the entire downstream transcriptional network. Glucose uptake and lipid storage can be reconstituted in MOF-depleted adipocytes in vitro by ectopic Glut4 expression, PPARγ agonist thiazolidinedione (TZD) treatment or SIRT1 inhibition. Hence, chronic imbalance in H4K16ac promotes a destabilisation of metabolism triggering the development of a metabolic disorder, and its maintenance provides an unprecedented regulatory epigenetic mechanism controlling diet-induced obesity.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
USP
Chloroquine phosphate, United States Pharmacopeia (USP) Reference Standard