Merck
  • EPB41L5 controls podocyte extracellular matrix assembly by adhesome-dependent force transmission.
EPB41L5 controls podocyte extracellular matrix assembly by adhesome-dependent force transmission.
Cell reports (2021-03-25)
Jasmin I Maier, Manuel Rogg, Martin Helmstädter, Alena Sammarco, Oliver Schilling, Benedikt Sabass, Jeffrey H Miner, Jörn Dengjel, Gerd Walz, Martin Werner, Tobias B Huber, Christoph Schell
ABSTRACT
The integrity of the kidney filtration barrier essentially relies on the balanced interplay of podocytes and the glomerular basement membrane (GBM). Here, we show by analysis of in vitro and in vivo models that a loss of the podocyte-specific FERM-domain protein EPB41L5 results in impaired extracellular matrix (ECM) assembly. By using quantitative proteomics analysis of the secretome and matrisome, we demonstrate a shift in ECM composition characterized by diminished deposition of core GBM components, such as LAMA5. Integrin adhesome proteomics reveals that EPB41L5 recruits PDLIM5 and ACTN4 to integrin adhesion complexes (IACs). Consecutively, EPB41L5 knockout podocytes show insufficient maturation of integrin adhesion sites, which translates into impaired force transmission and ECM assembly. These observations build the framework for a model in which EPB41L5 functions as a cell-type-specific regulator of the podocyte adhesome and controls a localized adaptive module in order to prevent podocyte detachment and thereby ensures GBM integrity.
Sigma-Aldrich
Anti-PDLIM5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ZYX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-LAMC1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Collagen Type IV α 2 Chain Antibody, clone 23IIC3, ascites fluid, clone 23IIC3, Chemicon®
Sigma-Aldrich
Anti-EPB41L5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution