Merck
  • Low-Cost Benzene Toluene Xylene Measurement Gas System Based on the Mini Chromatographic Cartridge.
Low-Cost Benzene Toluene Xylene Measurement Gas System Based on the Mini Chromatographic Cartridge.
Sensors (Basel, Switzerland) (2021-01-01)
Emiliano Zampetti, Paolo Papa, Joshua Avossa, Andrea Bearzotti, Catia Balducci, Giovanna Tranfo, Antonella Macagnano
ABSTRACT
Benzene, toluene and xylene (BTX) are an important part of the volatile organic compounds (VOCs) to be detected and monitored in the air, due to their toxicity towards human health. One of the most reliable technique used in BTX detection is gas chromatography (GC), which presents a high sensitivity. On the other hand, it has important drawbacks, such as high costs, the need for qualified personnel and frequent maintenance. To overcome these drawbacks, this work reports the development of a low cost and portable BTX gas detection system based on a mini chromatographic cartridge, a photo ionization detector (PID), a simple control unit (based on Arduino architecture) and a mini pump. In order to separate the BTX components, we propose the use of a cartridge 80 mm in length, composed of several commercial chromatographic column sections. To test the system performances, we have injected different amounts (from about 0.3 to 5.3 µg) of benzene, toluene and xylene and two of the most frequent possible interferents (ethanol, acetone). Experimental results have shown different retention time values (i.e., 25 ± 0.5 s, 51 ± 1.2 s and 117 ± 4 s, respectively) for benzene, toluene and xylene.
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%