Merck
  • Neoadjuvant Chemotherapy Shifts Breast Tumor Microbiota Populations to Regulate Drug Responsiveness and the Development of Metastasis.
Neoadjuvant Chemotherapy Shifts Breast Tumor Microbiota Populations to Regulate Drug Responsiveness and the Development of Metastasis.
Molecular cancer research : MCR (2019-10-20)
Akiko Chiba, Alaa Bawaneh, Christine Velazquez, Kenysha Y J Clear, Adam S Wilson, Marissa Howard-McNatt, Edward A Levine, Nicole Levi-Polyachenko, Shaina A Yates-Alston, Stephen P Diggle, David R Soto-Pantoja, Katherine L Cook
ABSTRACT
Breast tumors have their own specific microbiota, distinct from normal mammary gland tissue. Patients with breast cancer that present with locally advanced disease often undergo neoadjuvant chemotherapy to reduce tumor size prior to surgery to allow breast conservation or limit axillary lymph node dissection. The purpose of our study was to evaluate whether neoadjuvant chemotherapy modulates the tumor microbiome and the potential impact of microbes on breast cancer signaling. Using snap-frozen aseptically collected breast tumor tissue from women who underwent neoadjuvant chemotherapy (n = 15) or women with no prior therapy at time of surgery (n = 18), we performed 16S rRNA-sequencing to identify tumoral bacterial populations. We also stained breast tumor microarrays to confirm presence of identified microbiota. Using bacteria-conditioned media, we determined the effect of bacterial metabolites on breast cancer cell proliferation and doxorubicin therapy responsiveness. We show chemotherapy administration significantly increased breast tumor Pseudomonas spp. Primary breast tumors from patients who developed distant metastases displayed increased tumoral abundance of Brevundimonas and Staphylococcus. We confirmed presence of Pseudomonas in breast tumor tissue by IHC staining. Treatment of breast cancer cells with Pseudomonas aeruginosa conditioned media differentially effected proliferation in a dose-dependent manner and modulated doxorubicin-mediated cell death. Our results indicate chemotherapy shifts the breast tumor microbiome and specific microbes correlate with tumor recurrence. Further studies with a larger patient cohort may provide greater insights into the role of microbiota in therapeutic outcome and develop novel bacterial biomarkers that could predict distant metastases. IMPLICATIONS: Breast tumor microbiota are modified by therapy and affects molecular signaling.
Sigma-Aldrich
Phospholipase D from Streptomyces chromofuscus, ≥50,000 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Lipopolysaccharides from Pseudomonas aeruginosa 10, purified by phenol extraction
Sigma-Aldrich
Lectin from Pseudomonas aeruginosa, lyophilized powder
Sigma-Aldrich
Pyocyanin, Ready Made Solution from Pseudomonas aeruginosa, 5 mg/mL in DMSO
Sigma-Aldrich
Pseudomonas exotoxin A from Pseudomonas aeruginosa, lyophilized powder