Merck
  • Tailoring Hot Exciton Dynamics in 2D Hybrid Perovskites through Cation Modification.
Tailoring Hot Exciton Dynamics in 2D Hybrid Perovskites through Cation Modification.
ACS nano (2020-03-03)
Daniel B Straus, Sebastian Hurtado Parra, Natasha Iotov, Qinghua Zhao, Michael R Gau, Patrick J Carroll, James M Kikkawa, Cherie R Kagan
ABSTRACT
We report a family of two-dimensional hybrid perovskites (2DHPs) based on phenethylammonium lead iodide ((PEA)2PbI4) that show complex structure in their low-temperature excitonic absorption and photoluminescence (PL) spectra as well as hot exciton PL. We replace the 2-position (ortho) H on the phenyl group of the PEA cation with F, Cl, or Br to systematically increase the cation's cross-sectional area and mass and study changes in the excitonic structure. These single atom substitutions substantially change the observable number of and spacing between discrete resonances in the excitonic absorption and PL spectra and drastically increase the amount of hot exciton PL that violates Kasha's rule by over an order of magnitude. To fit the progressively larger cations, the inorganic framework distorts and is strained, reducing the Pb-I-Pb bond angles and increasing the 2DHP band gap. Correlation between the 2DHP structure and steady-state and time-resolved spectra suggests the complex structure of resonances arises from one or two manifolds of states, depending on the 2DHP Pb-I-Pb bond angle (as)symmetry, and the resonances within a manifold are regularly spaced with an energy separation that decreases as the mass of the cation increases. The uniform separation between resonances and the dynamics that show excitons can only relax to the next-lowest state are consistent with a vibronic progression caused by a vibrational mode on the cation. These results demonstrate that simple changes to the cation can be used to tailor the properties and dynamics of the confined excitons without directly modifying the inorganic framework.
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
2-Fluorophenethylamine, 99%
Sigma-Aldrich
2-Bromophenethylamine, 97%
Sigma-Aldrich
2-(2-Chlorophenyl)ethylamine, 95%