Skip to Content
We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.
Merck
Modulation of miR-21 signaling by MPS1 in human glioblastoma.
Oncotarget (2015-05-21)
Uday B Maachani, Anita Tandle, Uma Shankavaram, Tamalee Kramp, Kevin Camphausen
ABSTRACT
Monopolar spindle 1 (MPS1) is an essential spindle assembly checkpoint (SAC) kinase involved in determining spindle integrity. Beyond its mitotic functions, it has been implicated in several other signaling pathways. Our earlier studies have elaborated on role of MPS1 in glioblastoma (GBM) radiosensitization. In this study using reverse phase protein arrays (RPPAs), we assessed MPS1 mediated cell signaling pathways and demonstrated that inhibiting MPS1 could upregulate the expression of the tumor suppressor PDCD4 and MSH2 genes, by down regulating micro RNA-21 (miR-21). In GBMs miR-21 expression is significantly elevated and is associated with chemo and radioresistance. Both MPS1 and miR-21 depletion suppressed GBM cell proliferation, whereas, ectopic expression of miR-21 rescued GBM cell growth from MPS1 inhibition. Further, we demonstrate that MPS1 mediates phosphorylation of SMAD3 but not SMAD2 in GBM cells; A possible mechanism behind miR-21 modulation by MPS1. Collectively, our results shed light onto an important role of MPS1 in TGF-β/SMAD signaling via miR-21 regulation. We also, show the prognostic effect of miR-21, PDCD4 and MSH2 levels to patient survival across different GBM molecular subtypes. This scenario in which miR-21 is modulated by MPS1 inhibition may be exploited as a potential target for effective GBM therapy.
Sigma-Aldrich
Anti-Mps1 Antibody, CT, clone 4-112-3, clone 4-112-3, Upstate®, from mouse
Pricing and availability is not currently available. Contact Technical Service