Merck
  • Role of the AMPK/SREBP-1 pathway in the development of orotic acid-induced fatty liver.
Role of the AMPK/SREBP-1 pathway in the development of orotic acid-induced fatty liver.
Journal of lipid research (2011-07-16)
Eun-Jeong Jung, Sung-Won Kwon, Byung-Hwa Jung, Seon-Hee Oh, Byung-Hoon Lee
ABSTRACT
Orotic acid (OA), an intermediate in pyrimidine metabolism, has been used for a variety of purposes, such as dietary supplements. Although it is well documented that OA induces fatty liver in a species-specific manner, the precise molecular mechanisms remain unclear. The present study investigated the role of the adenosine monophosphate-activated protein kinase (AMPK)-sterol regulatory element-binding protein-1 (SREBP-1) pathway in the OA-induced fatty liver. Treatment with OA suppressed the phosphorylation of AMPK via proteasomal degradation of upstream kinase LKB1 and induced activation of SREBP-1 in both human hepatoma cell lines and primary rat hepatocytes. OA-induced SREBP-1 transcriptional activity was suppressed by cotreatment with aminoimidazole carboxamide ribonucleotide (AICAR) or metformin, or by overexpression of constitutively active AMPK (CA-AMPK) in the human hepatoma cell line. Importantly, in vivo data corroborated these results. Feeding 1% OA with diet decreased the phosphorylation of AMPK and increased the maturation of SREBP-1 and the expression of SREBP-responsive genes in the rat liver. OA-induced lipid accumulation was also completely inhibited by rapamycin. Mouse hepatocytes and mice were resistant to OA-induced lipogenesis because of little if any response in AMPK and downstream effectors. In conclusion, OA induces hepatic lipogenesis, mediated predominantly by the AMPK/SREBP-1 pathway in rat hepatocytes and human hepatoma cell lines.
Sigma-Aldrich
Orotic acid potassium salt, ≥98%
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous
Sigma-Aldrich
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranosyl 5′-monophosphate, ≥93%