Merck
  • DL-3-n-butylphthalide promotes dendrite development in cortical neurons subjected to oxygen-glucose deprivation/reperfusion.
DL-3-n-butylphthalide promotes dendrite development in cortical neurons subjected to oxygen-glucose deprivation/reperfusion.
Cell biology international (2018-04-27)
Peng Zhang, Ruxiang Xu, Yang Guo, Jiazhen Qin, Yiwu Dai, Ning Liu, Cuiying Wu
ABSTRACT
The limited degree of functional recovery is closely associated with the condition of the periinfarct cortex after ischemic stroke. The model of oxygen-glucose deprivation in cultured neurons could partly simulate this condition. Proper dendritic morphology is crucial for the correct wiring of neuronal function. Hence, the question of how to facilitate the plasticity of neural dendrites is of great significance. DL-3-n-butylphthalide is an efficient medication for ischemic stroke. In this study, in addition to having neuroprotective effects, DL-3-n-butylphthalide could increase the number of primary and secondary dendrites and of dendritic tips as confirmed by Sholl analysis. This study further demonstrated that DL-NBP inactivates PI3K/AKT signaling to positively regulate dendritic branching.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 15,000-30,000 by viscosity
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture