![Zinc stearate purum, 10-12% Zn basis](/deepweb/assets/sigmaaldrich/product/structures/176/333/6d12d4f8-2fb4-48a2-ac73-7fad21abeac2/640/6d12d4f8-2fb4-48a2-ac73-7fad21abeac2.png)
Zinc stearate
![Zinc stearate purum, 10-12% Zn basis](/deepweb/assets/sigmaaldrich/product/structures/176/333/6d12d4f8-2fb4-48a2-ac73-7fad21abeac2/640/6d12d4f8-2fb4-48a2-ac73-7fad21abeac2.png)
Synonym(s):
Stearic acid zinc salt
Linear Formula:
[CH3(CH2)16COO]2Zn
CAS Number:
Molecular Weight:
632.33
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing