Skip to Content
Merck
Vitamin D3(6,19,19-d3) solution 100 μg/mL in ethanol, 97 atom % D, 97% (CP)

Vitamin D3(6,19,19-d3) solution

Vitamin D3(6,19,19-d3) solution 100 μg/mL in ethanol, 97 atom % D, 97% (CP)
Synonym(s):
Calciol (6,19,19-d3) solution, Cholecalciferol (6,19,19-d3) solution
Empirical Formula (Hill Notation):
C27D3H41O
CAS Number:
Molecular Weight:
387.66
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing