
Vardenafil hydrochloride trihydrate

Empirical Formula (Hill Notation):
C23H32N6O4S · HCl · 3H2O
CAS Number:
Molecular Weight:
579.11
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing