
Trifluridine impurity A

Synonym(s):
5-Carboxy-2′-deoxyuridine
Empirical Formula (Hill Notation):
C10H12N2O7
CAS Number:
Molecular Weight:
272.21
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing