Skip to Content
Merck
trans-Platinum(II)diammine dichloride

trans-Platinum(II)diammine dichloride

trans-Platinum(II)diammine dichloride
Synonym(s):
trans-Diamminedichloroplatinum(II), trans-Dichlorodiammineplatinum(II)
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing